Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033560.3 | NP_291038.2 | 3 | 565 | ||
GenBank mRNA | AK016385.1 | - | 3 | 547 | ||
AK048942.1 | - | 3 | 545 | |||
AK054055.1 | - | 3 | 547 | |||
AK137479.1 | - | 3 | 550 | |||
AK157507.1 | - | 3 | 531 | |||
AK170280.1 | - | 3 | 534 | |||
AK170943.1 | - | 3 | 523 | |||
AK170961.1 | - | 3 | 554 | |||
AK171055.1 | - | 3 | 554 | |||
AK171221.1 | - | 3 | 566 | |||
BC039290.1 | - | 3 | 521 | 5066508 | ||
BC110396.1 | - | 3 | 480 | 5253683 |