Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080433.3 | NP_536681.2 | 2 | 404 | ||
GenBank mRNA | AB042399.1 | - | 1 | 121 | ||
AK019996.1 | - | 2 | 368 | |||
BC055718.1 | - | 1 | 731 | 6415359 | ||
BU703203.1 | - | 2 | 498 |