Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133673.3 | NP_598434.2 | 5 | 1392 | ||
GenBank mRNA | AK011416.1 | - | 1 | 120 | ||
AK028859.1 | - | 5 | 1206 | |||
AK035602.1 | - | 1 | 574 | |||
AK054404.1 | - | 5 | 1423 | |||
AK149754.1 | - | 5 | 1412 | |||
AK159843.1 | - | 5 | 1391 | |||
AK171756.1 | - | 3 | 2418 | |||
BC018456.1 | - | 5 | 1391 | 3582246 |