Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cherp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138585.3 | NP_613051.3 | 2 | 237 | ||
GenBank mRNA | AF536541.1 | - | 2 | 139 | ||
AK017525.1 | - | 2 | 168 | |||
AK049765.1 | - | 2 | 146 | |||
AK144328.1 | - | 2 | 122 | |||
BC032191.1 | - | 2 | 128 | 5322208 | ||
BY061354.1 | - | 2 | 238 |