Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138589.2 | NP_613055.2 | 2 | 554 | ||
GenBank mRNA | AK011862.1 | - | 2 | 310 | ||
AK017851.1 | - | 2 | 331 | |||
AK033428.1 | - | 2 | 546 | |||
AK077395.1 | - | 2 | 577 | |||
AK135258.1 | - | 2 | 312 | |||
AK140891.1 | - | 2 | 313 | |||
BC011313.1 | - | 2 | 557 | 3499738 |