Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006834.4 | NP_006825.1 | 2 | 479 | ||
GenBank mRNA | AF498958.1 | - | 2 | 299 | ||
AK312202.1 | - | 2 | 338 | |||
BC015061.1 | - | 2 | 481 | 4280407 | ||
U59878.1 | - | 2 | 254 | |||
U71127.1 | - | 2 | 312 |