Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290792.1 | NP_001277721.1 | 8 | 823 | ||
NM_001290794.1 | NP_001277723.1 | 8 | 719 | |||
NM_001290795.1 | NP_001277724.1 | 8 | 823 | |||
NM_172372.2 | NP_758960.1 | 8 | 819 | |||
GenBank mRNA | AF229635.1 | - | 8 | 797 | ||
AK018409.1 | - | 8 | 818 | |||
AK145616.1 | - | 8 | 719 | |||
AK208276.1 | - | 3 | 284 | |||
BC011479.1 | - | 8 | 790 | 4035468 |