Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172631.3 | NP_766219.2 | 3 | 669 | ||
GenBank mRNA | AK052338.1 | - | 3 | 670 | ||
BC096371.1 | - | 2 | 430 | 3583946 | ||
BC131933.1 | - | 2 | 277 | 40130210 | ||
BC131935.1 | - | 2 | 277 | 40130212 |