Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177470.3 | NP_803421.1 | 2 | 135 | ||
GenBank mRNA | AK002555.1 | - | 2 | 125 | ||
AK050101.1 | - | 2 | 137 | |||
AK167567.1 | - | 2 | 136 | |||
AK167715.1 | - | 2 | 133 | |||
AK169359.1 | - | 2 | 136 | |||
BC028901.1 | - | 2 | 130 | 4195488 |