Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lair1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178611.6 | NP_848726.1 | 2, 3 | 176 | ||
GenBank mRNA | AF491830.1 | - | 2, 3 | 88 | ||
AK017222.1 | - | 2, 3 | 180 | |||
AK033472.1 | - | 2, 3 | 153 | |||
AK038574.1 | - | 2, 3 | 139 | |||
AY392764.1 | - | 2, 3 | 113 |