Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008928.4 | NP_032954.1 | 12 | 1274 | ||
GenBank mRNA | AK008141.1 | - | 12 | 1277 | ||
AK011002.1 | - | 12 | 1275 | |||
AK150067.1 | - | 12 | 1275 | |||
AK159379.1 | - | 12 | 1273 | |||
AK166255.1 | - | 12 | 1276 | |||
AK166507.1 | - | 12 | 1275 | |||
AK167209.1 | - | 12 | 1275 | |||
AK167442.1 | - | 12 | 1278 | |||
AK168710.1 | - | 12 | 1257 | |||
AK176968.1 | - | 3 | 244 | |||
BC007467.1 | - | 11 | 1213 | 3497714 |