Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081567.1 | NP_001075036.1 | 4 | 744 | ||
NM_009158.2 | NP_033184.2 | 4 | 744 | |||
GenBank mRNA | AB005665.1 | - | 1 | 127 | ||
AB096076.1 | - | 2 | 178 | |||
AB096077.1 | - | 2 | 178 | |||
AB096078.1 | - | 2 | 178 | |||
AB096079.1 | - | 2 | 178 | |||
AK036275.1 | - | 4 | 696 | |||
AK042368.1 | - | 3 | 309 | |||
AK082242.1 | - | 4 | 747 | |||
AK163206.1 | - | 4 | 695 | |||
BC026697.1 | - | 4 | 736 | |||
BC046625.1 | - | 4 | 683 | 6494163 | ||
L35236.1 | - | 4 | 487 |