Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009326.2 | NP_033352.1 | 3 | 294 | ||
GenBank mRNA | AB010271.1 | - | 3 | 232 | ||
AB221557.1 | - | 3 | 197 | |||
AK131650.1 | - | 3 | 551 | |||
AK137527.1 | - | 3 | 176 | |||
BC049617.1 | - | 3 | 287 | 6773546 | ||
BC065786.1 | - | 3 | 285 | 5120316 | ||
BC138504.1 | - | 3 | 283 | 8861524 | ||
BC138505.1 | - | 3 | 283 | 8861525 | ||
D86081.1 | - | 3 | 294 |