Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009327.3 | NP_033353.2 | 10 | 2059 | ||
GenBank mRNA | AK141225.1 | - | 10 | 1865 | ||
AK144051.1 | - | 10 | 2067 | |||
BC080698.1 | - | 10 | 2061 | 30471380 | ||
M57966.1 | - | 2039 |