Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp354a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009329.3 | NP_033355.2 | 5 | 1937 | ||
GenBank mRNA | AF184111.1 | - | 5 | 1899 | ||
AK030469.1 | - | 5 | 1916 | |||
AK083769.1 | - | 1 | 132 | |||
BC050843.1 | - | 5 | 1937 | 6506256 | ||
BC087540.1 | - | 5 | 2073 | 4239789 | ||
L77247.1 | - | 4 | 1801 |