Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009331.3 | NP_033357.1 | 9 | 852 | ||
GenBank mRNA | AK037616.1 | - | 9 | 853 | ||
AK038140.1 | - | 3 | 190 | |||
BC056630.1 | - | 8 | 1094 | |||
BC138596.1 | - | 8 | 832 | 8861616 | ||
BC145343.1 | - | 8 | 832 | 9053899 | ||
X61385.1 | - | 10 | 994 |