Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 4 | 603 | ||
GenBank mRNA | AB053464.1 | - | 4 | 548 | ||
AF316616.2 | - | 4 | 460 | |||
AK154050.1 | - | 4 | 579 | |||
AK160587.1 | - | 4 | 544 | |||
AK194210.1 | - | 1 | 145 | |||
AK197443.1 | - | 1 | 145 | |||
AY338234.1 | - | 4 | 533 | |||
BC010716.1 | - | 4 | 516 | 2655861 |