Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009464.3 | NP_033490.1 | 4 | 674 | ||
GenBank mRNA | AB008216.1 | - | 3 | 477 | ||
AB010742.1 | - | 4 | 674 | |||
AB013132.1 | - | 2 | 230 | |||
AF030164.1 | - | 3 | 515 | |||
AF032902.1 | - | 4 | 641 | |||
AF053352.1 | - | 4 | 646 | |||
BC055901.1 | - | 3 | 864 | |||
BC098234.1 | - | 2 | 1370 | |||
BC139430.1 | - | 3 | 542 | 8862452 | ||
BC139431.1 | - | 3 | 542 | 8862453 |