Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009464.3 | NP_033490.1 | 6, 7 | 1016 | ||
GenBank mRNA | AB008216.1 | - | 5, 6 | 819 | ||
AB010742.1 | - | 6, 7 | 1016 | |||
AF030164.1 | - | 5, 6 | 857 | |||
AF032902.1 | - | 6, 7 | 983 | |||
AF053352.1 | - | 6, 7 | 988 | |||
BC055901.1 | - | 5, 6 | 1206 | |||
BC098234.1 | - | 4, 5 | 1712 | |||
BC139430.1 | - | 5, 6 | 884 | 8862452 | ||
BC139431.1 | - | 5, 6 | 884 | 8862453 |