Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009481.2 | NP_033507.2 | 7 | 1116 | ||
GenBank mRNA | AK028443.1 | - | 7 | 1101 | ||
AK049283.1 | - | 8 | 1229 | |||
DQ086491.1 | - | 7 | 1077 | |||
U67874.1 | - | 7 | 1077 |