Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009502.4 | NP_033528.3 | 1 | 162 | ||
GenBank mRNA | AK077850.1 | - | 1 | 198 | ||
AK135530.1 | - | 1 | 198 | |||
BC008520.1 | - | 1 | 162 | 3495710 | ||
L18880.1 | - | 1 | 91 |