Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009560.2 | NP_033586.2 | 5 | 717 | ||
NM_029531.2 | NP_083807.2 | 5 | 523 | |||
GenBank mRNA | AK028717.1 | - | 5 | 617 | ||
AK149305.1 | - | 5 | 544 | |||
BC031754.1 | - | 5 | 522 | 3992720 | ||
CB526626.1 | - | 5 | 660 |