Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011614.3 | NP_035744.1 | 7 | 1365 | ||
GenBank mRNA | AF030100.2 | - | 7 | 1018 | ||
AK020909.1 | - | 4 | 704 | |||
AK170297.1 | - | 7 | 1265 | |||
AK171014.1 | - | 7 | 1265 | |||
CB848966.2 | - | 1 | 139 |