Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136259.1 | NP_001129731.1 | 16 | 1900 | ||
NM_011622.3 | NP_035752.1 | 15 | 1828 | |||
GenBank mRNA | AJ006972.1 | - | 15 | 1805 | ||
AK028398.1 | - | 15 | 1769 | |||
AK051245.1 | - | 15 | 1810 | |||
AK160403.1 | - | 15 | 1774 | |||
AK203247.1 | - | 1 | 156 | |||
BC093520.1 | - | 15 | 1756 | 2866013 | ||
BC157905.1 | - | 15 | 1789 | 9088106 | ||
BC158042.1 | - | 15 | 1789 | 9007343 |