Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011624.2 | NP_035754.1 | 7, 8 | 1020 | ||
GenBank mRNA | AB013603.1 | - | 6, 7 | 833 | ||
AB045324.1 | - | 7, 8 | 1094 | |||
AK041159.1 | - | 7, 8 | 930 | |||
AK144709.1 | - | 8, 9 | 1006 | |||
AK146475.1 | - | 7, 8 | 1042 | |||
AK205914.1 | - | 13 | ||||
BC031723.1 | - | 7, 8 | 1020 | 5003002 |