Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177701.2 | NP_001171172.1 | 3 | 555 | ||
NM_006860.4 | NP_006851.1 | 3 | 552 | |||
XM_006724106.2 | XP_006724169.1 | 4 | 316 | |||
XM_017028540.1 | XP_016884029.1 | 3 | 559 | |||
GenBank mRNA | AB209424.1 | - | 3 | 547 | ||
AK307105.1 | - | 3 | 515 | |||
AK307382.1 | - | 3 | 524 | |||
AL022729.1 | - | 3 | 496 | |||
BC000566.2 | - | 3 | 533 | 3163341 | ||
BM711121.1 | - | 3 | 143 | |||
BT006815.1 | - | 3 | 129 | |||
CR456558.1 | - | 3 | 171 |