Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011667.2 | NP_035797.1 | 5 | 485 | ||
GenBank mRNA | AF150963.1 | - | 5 | 485 | ||
AK030031.1 | - | 5 | 484 |