Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005201.3 | NP_005192.1 | 2 | 575 | ||
GenBank mRNA | AK314957.1 | - | 2 | 577 | ||
BC069067.1 | - | 1 | 448 | 7216880 | ||
BC074743.2 | - | 2 | 477 | 30915201 | ||
BC107152.2 | - | 3 | 335 | 40032486 | ||
BC107159.1 | - | 2 | 508 | 40032938 | ||
D49919.1 | - | 2 | 508 |