Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011675.2 | NP_035805.2 | 3 | 416 | ||
GenBank mRNA | BC025146.1 | - | 3 | 383 | 4913412 | |
L31783.1 | - | 3 | 276 |