Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011678.2 | NP_035808.2 | 3, 4 | 440 | ||
GenBank mRNA | AK143582.1 | - | 3, 4 | 375 | ||
AK149964.1 | - | 3, 4 | 398 | |||
AK169372.1 | - | 3, 4 | 419 | |||
AK169933.1 | - | 3, 4 | 410 | |||
AK171271.1 | - | 3, 4 | 410 | |||
AK171633.1 | - | 3, 4 | 393 | |||
AK214431.1 | - | 2, 3 | 236 | |||
AK218565.1 | - | 2, 3 | 236 | |||
AK220512.1 | - | 3, 4 | 397 | |||
BC011341.1 | - | 3, 4 | 358 | 3661165 | ||
BC066180.1 | - | 3, 4 | 350 | |||
BC066865.1 | - | 3, 4 | 350 | |||
L00681.1 | - | 3, 4 | 403 |