Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130917.2 | NP_001124389.2 | 3 | 218 | ||
NM_001290270.1 | NP_001277199.1 | 2 | 182 | |||
NM_001290271.1 | NP_001277200.1 | 4 | 657 | |||
NM_006866.3 | NP_006857.2 | 4 | 790 | |||
XM_006722986.1 | XP_006723049.1 | 4 | 790 | |||
XM_011526385.1 | XP_011524687.1 | 4 | 790 | |||
XM_011526386.1 | XP_011524688.1 | 3 | 754 | |||
XM_011526387.1 | XP_011524689.1 | 3 | 754 | |||
XM_011526388.1 | XP_011524690.1 | 3 | 754 | |||
XM_011526390.1 | XP_011524692.1 | 4 | 790 | |||
XM_011526391.1 | XP_011524693.1 | 4 | 790 | |||
XM_017026224.1 | XP_016881713.1 | 4 | 790 | |||
GenBank mRNA | AF025531.1 | - | 3 | 209 | ||
AK130170.1 | - | 3 | 248 | |||
AK222651.1 | - | 3 | 214 | |||
AK310088.1 | - | 4 | 639 | |||
BC017412.1 | - | 3 | 220 | 4618777 | ||
BC027916.1 | - | 3 | 203 | 5223154 | ||
BP296918.1 | - | 3 | 259 | |||
EU915609.1 | - | 2 | 151 | |||
EU915610.1 | - | 3 | 187 | |||
U70665.1 | - | 3 | 219 | |||
U82275.1 | - | 3 | 257 | |||
U82276.1 | - | 2 | 172 | |||
U82277.1 | - | 3 | 170 | |||
U82278.1 | - | 3 | 170 |