siRNA Details
Gene Symbol: Vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011694.4 | NP_035824.1 | 8, 9 | 798 | ||
GenBank mRNA | AK002740.1 | - | 8, 9 | 857 | ||
AK146239.1 | - | 8, 9 | 831 | |||
AK153094.1 | - | 8, 9 | 800 | |||
AK167860.1 | - | 8, 9 | 831 | |||
AK168225.1 | - | 8, 9 | 807 | |||
AK168672.1 | - | 8, 9 | 836 | |||
AK169160.1 | - | 8, 9 | 836 | |||
AK169282.1 | - | 8, 9 | 840 | |||
AK169354.1 | - | 8, 9 | 836 | |||
AK169671.1 | - | 8, 9 | 797 | |||
BC092257.1 | - | 8, 9 | 781 | 5124735 | ||
U30840.1 | - | 8, 9 | 798 |

More Information
Other Species Targeted

Panther Classification:
