Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011715.2 | NP_035845.1 | 3 | 274 | ||
GenBank mRNA | AF020055.1 | - | 3 | 274 | ||
AK004644.1 | - | 3 | 382 | |||
AK004858.1 | - | 3 | 345 | |||
AK151146.1 | - | 3 | 273 | |||
AK153338.1 | - | 3 | 285 | |||
AK156300.1 | - | 3 | 311 | |||
AK165225.1 | - | 3 | 244 | |||
AK165323.1 | - | 3 | 244 | |||
AK167241.1 | - | 3 | 300 | |||
AK167275.1 | - | 3 | 342 | |||
AK167813.1 | - | 3 | 272 | |||
AK169835.1 | - | 3 | 368 | |||
AK169893.1 | - | 3 | 304 | |||
BC049117.1 | - | 3 | 381 | |||
BC119358.1 | - | 3 | 230 | 8734107 | ||
BC125295.1 | - | 3 | 230 | 40129810 |