Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011718.2 | NP_035848.1 | 5 | 2150 | ||
GenBank mRNA | AK029284.1 | - | 4 | 1593 | ||
U30464.1 | - | 5 | 2148 |