Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116258.1 | - | 3 | 448 | 40093733 | |
BC116259.1 | - | 3 | 448 | 40093735 | ||
Y14695.1 | - | 3 | 449 |