Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011794.3 | NP_035924.2 | 6 | 573 | ||
GenBank mRNA | AF125043.1 | - | 6 | 531 | ||
AK008091.1 | - | 5 | 426 | |||
AK008714.1 | - | 6 | 554 | |||
AK093938.1 | - | 6 | 559 | |||
AK150877.1 | - | 6 | 553 | |||
AK151931.1 | - | 6 | 577 | |||
AK152009.1 | - | 6 | 553 | |||
AK154435.1 | - | 6 | 553 | |||
BC011036.1 | - | 6 | 526 | 4160742 | ||
BY744642.1 | - | 6 | 573 |