Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 4 | 445 | ||
NM_011806.3 | NP_035936.3 | 4 | 445 | |||
GenBank mRNA | AK029147.1 | - | 4 | 387 | ||
AK030508.1 | - | 4 | 414 | |||
AK033208.1 | - | 4 | 411 | |||
AK039563.1 | - | 4 | 413 | |||
AK040107.1 | - | 4 | 403 | |||
AK046201.1 | - | 4 | 341 | |||
AK078015.1 | - | 4 | 346 | |||
AK145803.1 | - | 4 | 407 | |||
AK160595.1 | - | 4 | 418 | |||
BC060109.1 | - | 4 | 368 | |||
U70017.1 | - | 4 | 400 |