Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145937.1 | NP_001139409.1 | 21 | 4146 | ||
NM_011857.3 | NP_035987.3 | 23 | 4194 | |||
GenBank mRNA | AB025412.1 | - | 23 | 4194 | ||
AF195418.1 | - | 17 | 3039 | |||
AK122513.1 | - | 9 | 1485 | |||
AK135844.1 | - | 9 | 1617 | |||
BC145284.1 | - | 21 | 4140 | 9053838 |