Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011875.4 | NP_036005.1 | 7 | 590 | ||
GenBank mRNA | AF107838.1 | - | 7 | 551 | ||
AK051078.1 | - | 7 | 581 | |||
AK075936.1 | - | 7 | 591 | |||
AK081160.1 | - | 7 | 588 | |||
AK145654.1 | - | 7 | 553 | |||
AK163017.1 | - | 7 | 585 | |||
AK167169.1 | - | 7 | 575 | |||
AK171504.1 | - | 7 | 571 | |||
AK193895.1 | - | 1 | 65 | |||
BC029989.1 | - | 7 | 530 | |||
BC139184.1 | - | 7 | 568 | 8862206 | ||
BC139187.1 | - | 7 | 568 | 8862209 |