Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011882.2 | NP_036012.1 | 4 | 753 | ||
GenBank mRNA | AF281045.1 | - | 2 | 297 | ||
AK053125.1 | - | 3 | 451 | |||
AK054205.1 | - | 2 | 1248 | |||
AK140815.1 | - | 4 | 754 | |||
BC003873.1 | - | 4 | 562 | |||
BC138013.1 | - | 1 | 170 | 8861032 | ||
JX443419.1 | - | 1 | 134 |