Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF037205.1 | - | 6 | 559 | ||
AF037206.1 | - | 6 | 559 | |||
AK034135.1 | - | 6 | 620 | |||
AK036591.1 | - | 6 | 604 | |||
AK048381.1 | - | 1 | 1705 | |||
AK050095.1 | - | 6 | 1304 | |||
AK082296.1 | - | 6 | 600 | |||
AK139447.1 | - | 6 | 565 | |||
AK140682.1 | - | 6 | 614 | |||
AK158046.1 | - | 6 | 600 | |||
AK167278.1 | - | 6 | 564 | |||
BC058182.1 | - | 6 | 567 | 5324040 |