Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011895.3 | NP_036025.2 | 6 | 915 | ||
GenBank mRNA | AK144170.1 | - | 6 | 908 | ||
BC012252.1 | - | 6 | 830 | 4190458 | ||
Z71268.1 | - | 6 | 736 |