Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp54c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100110.1 | NP_001093580.1 | 903 | |||
GenBank mRNA | AK090018.1 | - | 906 | |||
AK146826.1 | - | 942 | ||||
BC005543.1 | - | 920 | 3585216 |
Gene Symbol: Srp54a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011899.4 | NP_036029.2 | 9 | 977 | ||
GenBank mRNA | AK011928.1 | - | 9 | 941 | ||
BC019683.1 | - | 9 | 906 | 3708250 |
Gene Symbol: Srp54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100109.1 | NP_001093579.1 | 9 | 1213 | ||
GenBank mRNA | AK136033.1 | - | 9 | 1214 |