Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011932.2 | NP_036062.1 | 7 | 781 | ||
GenBank mRNA | AF163255.1 | - | 7 | 735 | ||
AF186023.1 | - | 7 | 756 | |||
AK076886.1 | - | 5 | 334 | |||
AK145745.1 | - | 7 | 747 | |||
AK146180.1 | - | 7 | 750 | |||
AK169584.1 | - | 6 | 646 | |||
BC014759.1 | - | 7 | 777 | 4486834 |