Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159500.1 | NP_001152972.1 | 2 | 667 | ||
NM_011934.4 | NP_036064.3 | 2 | 566 | |||
GenBank mRNA | AK032025.1 | - | 2 | 374 | ||
AK044339.1 | - | 2 | 567 | |||
AK052256.1 | - | 2 | 669 | |||
BC044858.1 | - | 1 | 236 | |||
BC132595.1 | - | 2 | 531 | 40130872 | ||
BC132597.1 | - | 2 | 532 | 40130874 | ||
S82458.1 | - | 1 | 274 | |||
X89594.1 | - | 1 | 274 |