Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038018.4 | NP_001033107.1 | 15 | 1833 | ||
NM_001112711.2 | NP_001106182.1 | 15 | 1833 | |||
NM_001286063.1 | NP_001272992.1 | 15 | 1813 | |||
NM_001286064.1 | NP_001272993.1 | 15 | 1818 | |||
NM_001286065.1 | NP_001272994.1 | 15 | 1731 | |||
NM_001286066.1 | NP_001272995.1 | 15 | 1826 | |||
NM_011938.4 | NP_036068.2 | 15 | 1833 | |||
GenBank mRNA | AF040747.1 | - | 15 | 1703 | ||
AF040748.1 | - | 15 | 1703 | |||
AF040749.1 | - | 15 | 1703 | |||
AK051405.1 | - | 15 | 1815 | |||
AK146119.1 | - | 15 | 1606 | |||
AK161142.1 | - | 13 | 1465 | |||
AK180303.1 | - | 3 | 294 | |||
BC057206.1 | - | 15 | 1806 | |||
BC075719.1 | - | 15 | 1714 | 5031455 | ||
CA528447.1 | - | 4 | 432 | |||
CD354984.1 | - | 2 | 173 | |||
Y15797.1 | - | 15 | 1655 | |||
Y15798.1 | - | 15 | 1677 | |||
Y15799.1 | - | 15 | 1660 | |||
Y15800.1 | - | 15 | 1648 |