Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011956.3 | NP_036086.1 | 3 | 321 | ||
GenBank mRNA | AF114169.1 | - | 3 | 299 | ||
AK003816.1 | - | 3 | 319 | |||
AK169896.1 | - | 3 | 280 | |||
AK210395.1 | - | 1 | 105 | |||
AK213247.1 | - | 1 | 104 | |||
BC012635.1 | - | 3 | 322 | 4038123 | ||
BI735707.1 | - | 3 | 321 |