Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011967.3 | NP_036097.1 | 8 | 986 | ||
GenBank mRNA | AF019661.1 | - | 8 | 701 | ||
AK143468.1 | - | 8 | 988 | |||
AK160530.1 | - | 8 | 746 | |||
AK160550.1 | - | 8 | 740 | |||
AK160741.1 | - | 8 | 749 | |||
AK161680.1 | - | 8 | 734 | |||
BC007181.1 | - | 8 | 733 | |||
BC010709.1 | - | 8 | 722 | 2648635 | ||
BC083342.1 | - | 8 | 719 | 5700963 | ||
BC108368.1 | - | 8 | 716 | 3596530 |