Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011979.2 | NP_036109.3 | 3 | 562 | ||
GenBank mRNA | AJ132103.1 | - | 3 | 552 | ||
BC111521.2 | - | 3 | 534 | 40047284 |