Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110796.2 | NP_001104266.1 | 3 | 2563 | ||
NM_011995.4 | NP_036125.4 | 3 | 2563 | |||
GenBank mRNA | AB083477.1 | - | 3 | 2488 | ||
AB083478.1 | - | 3 | 2488 | |||
Y19185.2 | - | 4 | 2473 | |||
Y19186.2 | - | 4 | 2473 |